Learn More
American Research Products Inc Rat Runt related transcription
Supplier:  American Research Products Inc CSBEL020594RA
Encompass
Rat Runt-related transcription factor 2 (RUNX2) ELISA kit from CUSABIO Detection Range 3 12 ng/mL-200 ng/mL Sensitivity 0 78 ng/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-27152