Learn More
American Research Products Inc Rat secretory immunoglobulin A
Supplier:  American Research Products Inc CSBE08412R
Encompass
Rat secretory immunoglobulin A sIgA ELISA Kit from CUSABIO Detection Range 0 78 ng/mL-50 ng/mL Sensitivity 0 195 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-30212