Learn More
BIOHIPPO Rat TARC Microsample
SDP
Supplier:  BIOHIPPO BHE15211243
Encompass
Rat TARC Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TARC in Serum plasma tissue homogenates cell lysates and other biological fluids It provides a detection range of 15 63-1000 pg/mL with a sensitivity of 6 1 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat TARC Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat TARC Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat TARC biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14554