Learn More
Innovative Research Inc Rat Testis Tissue Lysate 100ug
Supplier:  Innovative Research Inc IRTTSTTL100UG
Encompass_Preferred
Rat Testis Tissue Lysate 100ug - Rat Testis Tissue Lysate from Innovative Research is provided as a Liquid buffered in SDS sample buffer containing 5% beta-mercaptoethanol. This product is especially useful for applications like Western Blot, among others. For continuous use, store at 2-8C. If not using the entire volume immediately, store single-use aliquots at -70C. Repeated freeze/thaw cycles are not recommended. Under proper storage conditions, the shelf life is six months from the date of receipt. Additional Details Species Rat Source Testis tissue lysate Format Liquid Storage Conditions -70C
Catalog No. 50-274-3990