Learn More
American Research Products Inc Rat Testosterone T ELISA Kit
Supplier:  American Research Products Inc CSBE05100R
Encompass
Rat Testosterone T ELISA Kit from CUSABIO Detection Range 0 13 ng/mL-25 6 ng/mL Sensitivity 0 06 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Competitive
Catalog No. 50-003-26828