Learn More
American Research Products Inc Rat thrombin antithrombin comp
Supplier:  American Research Products Inc CSBE08432R
Encompass
Rat thrombin-antithrombin compleX TAT ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma cell culture supernates tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-26693