Learn More
American Research Products Inc Rat thyroXine T4 ELISA Kit
Supplier:  American Research Products Inc CSBE05082R
Encompass
Rat thyroXine T4 ELISA Kit from CUSABIO Detection Range 20 ng/mL-320 ng/mL Sensitivity 20 ng/mL Sample serum plasma cell culture supernates tissue homogenates cell lysates Method Competitive
Catalog No. 50-003-26829