Learn More
American Research Products Inc Rat Tri iodothyronine T3 ELIS
Supplier:  American Research Products Inc CSBE05085R
Encompass
Rat Tri-iodothyronine T3 ELISA Kit from CUSABIO Detection Range 0 5 ng/mL-8 ng/mL Sensitivity 0 5 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Competitive
Catalog No. 50-003-29132