Learn More
Instech Rat Vascular Access Button tether kit for ABS: 10.5in VABR1T/25 (white port), 375/25PS, 9in 015 silicone
SDP
Supplier:  Instech KVABR1T/25ABS
Encompass
The Instech KVABR1T/25-ABS rat tether kit is designed for sampling from the white port of a one-channel VAB™ (VABR1B/22). It includes a 10.5in (27cm) tether (VABR1T/25), a 25ga plastic swivel (375/25PS), 9in (23cm) of 0.015in ID silicone tubing, and 13.5in (34cm) of PU tubing. A 10.5in (27cm) PS95 spring provides strain relief, and the 25ga connector mates exclusively with the white port. Configured for ABS2™ automated blood sampling workflows.
Features
  • 1-channel white-port rat VAB™ tether kit
  • 10.5in (27cm) VABR1T/25 tether
  • 375/25PS swivel, 25ga
  • 9in (23cm) 0.015in silicone + 13.5in (34cm) PU tubing
  • 10.5in (27cm) PS95 spring
  • ABS2-compatible; white port only
Catalog No. NC1322390
 Restricted Item