Learn More
Cusabio Technology LLC Rat visfatin ELISA Kit
Supplier:  Cusabio Technology LLC CSBE08941R
Encompass_Preferred
The Rat Visfatin ELISA Kit is designed for the quantitative detection of NAMPT (visfatin) in serum plasma and tissue homogenates using a sandwich assay format This kit detects NAMPT within a broad dynamic range of 3 12 ng/mL to 200 ng/mL with a high sensitivity of 0 78 ng/mL ensuring precise measurement of low-abundance samples The assay requires a short incubation time of 1-5 hours and a modest sample volume of 50-100 L per test Detection is performed at 450 nm providing reliable and reproducible results for research applications Its compatibility with multiple biological matrices makes it a versatile tool for studying NAMPT levels in metabolic and inflammatory research
Catalog No. 50-000-49362