Learn More
Lonza Walkersville Rat Wistar Heps Plateable
Supplier: Lonza Walkersville RICP01
All our plateable cryopreserved primary animal hepatocytes are specifically pre tested for metabolic activity of general Phase I and Phase II metabolism in suspension They are tested for healthy morphology lasting 72 hours in monlayer sandwich culture