Learn More
Medchemexpress LLC Raxatrigine | 934240-30-9 | 99.9% | 10 MG

Supplier: Medchemexpress LLC HY1279610MG
Raxatrigine is a novel small molecule state-dependent sodium channel blocker and Nav1.7 sodium channel inhibitor. It is for research use only.
- Purity of 99.85%.
- CAS number 934240-30-9.
- Molecular weight of 314.35.
- Chemical formula C18H19FN2O2.
- Appears as a white to off-white solid.
- Inhibits Nav1.7 sodium channels.
- Received orphan-drug designation from the US Food and Drug Administration in July 2013.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.