Learn More
Medchemexpress LLC Raxatrigine (Vixotrigine) | 934240-30-9 | 99.9% | 50 MG

Supplier: Medchemexpress LLC HY1279650MG
Raxatrigine (Vixotrigine) is a novel small molecule state-dependent sodium channel blocker and a Nav1.7 sodium channel inhibitor. This compound is intended for research use only.
- Nav1.7 sodium channel inhibitor
- Available as a white to off-white solid
- Ships at room temperature in the continental US
- Store powder at -20°C for 3 years or 4°C for 2 years
- Store in solvent at -80°C for 6 months or -20°C for 1 month
- Purity of 99.9%
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.