Learn More
Raybiotech Inc RayBiotech Cell Lysate, HeLa, UV Treated, Lyophilized, 1 vial
Supplier: Raybiotech Inc HELAUVS0011
HeLa Cell Lysate, UV Treated. HeLa cells were treated with UV and solubilized at 4 x 106 cells/100μl in cell lysis buffer.
- Size: 1 vial
- Format: Lyophilized
- Recommended applications: SDS-PAGE, IP, WB, Sandwich ELISA, Array
- Shipping type: Blue ice
- Storage: -20°C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.