Learn More
Medchemexpress LLC Razpipadon (racemate) | 1643462-93-4 | 97.1% | C19H17F2N3O4 | 10MG
SDP
Supplier:  Medchemexpress LLC HY14149510MG
Encompass_Preferred
(Rac)-Razpipadon is a research-grade noncatechol dopamine D1 receptor agonist and G protein-biased ligand supplied for in vitro pharmacology and signaling studies. It is provided as a white to off-white solid and is characterized by nanomolar potency in cAMP assays, supporting use in receptor pharmacology and mechanism-of-action experiments.
  • Nanomolar potency (EC50 5.8 nM) in Gs-cAMP assays.
  • G protein-biased D1 receptor agonist for signaling studies.
  • Noncatechol scaffold useful for mechanism and selectivity investigations.
  • Supplied as a white to off-white solid for laboratory handling.
  • High supplied purity (97.09%) for reproducible results.
  • Available in multiple research sizes and solution forms for rapid use.
Catalog No. 50-002-61429