Learn More
Cytek Biosciences Inc Rb aGt IgG Anti Secondary
Supplier: Cytek Biosciences Inc 728095M001
Product Name: HRP Rabbit Anti-Goat IgG (H&L)
Product Class: Antibody
Type: Secondary
Product Name: HRP Rabbit Anti-Goat IgG (H&L)
Cytek Catalog Number: 72-8095-M001
Marker: IgG (H&L)
Target Species: Goat
Reported Cross Reactivity: Goat
Clonality: Polyclonal
Host Species: Rabbit
Conjugation: HRP
Size: 1 mg
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.