Learn More
Foxx Life Sciences Borosil® Round Bottom Boiling Flask with Interchangeable Joint 24/29, 100mL, 20/CS
Supplier:  Foxx Life Sciences 4380C16
Encompass_Preferred
Borosil® Round Bottom Boiling Flasks have uniform wall thickness for even heat distribution. Constructed from stress-free borosilicate. Borosil® Boiling Flasks are heat-resistant. Built to withstand the demands of the modern lab, each flask has a flame-polished, Interchangable Joint, and an enamel area for marking. Available from 25mL to 4L. Flasks comply with ISO 4797. Some options of 25mL, 50mL, 500mL, 1L and 2L options not covered in ISO. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil® Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity, and reliability at competitive prices. And thanks to a unique annealing process, Borosil® Glassware is stress-free and incredibly resistant to thermal shock.
Catalog No. 50-172-0503