Learn More
Biocare Medical, LLC Rabbit-on-Rodent HRP-Polymer, 25ml
SDP
Supplier:  Biocare Medical, LLC RMR622H
Encompass_Preferred
The Rabbit-on-Rodent HRP-Polymer is for use of rabbit primary antibodies on mouse or rat tissues. Rabbit antibodies display minimum cross-reactivity to endogenous rodent IgG. It is labeled with horseradish peroxidase (HRP).
Catalog No. 50-832-79