Learn More
BD Cell Analysis 3P BD OptiBuild™ RB670 Mouse Anti-Human EphB2, 50 µg
Supplier:  BD Cell Analysis 3P 770103
Encompass
Brand: BD OptiBuild™, Alternative Name: EPHB2; EphB2R; TYRO5; CAPB; DRT; EK5; EPHT3; EPTH3; HEK5; PCBC, Reactivity: Human (Tested in Development), Isotype: Mouse BALB/c IgG1, κ, Immunogen: Human EphB2 Recombinant Protein, Application: Flow cytometry (Qualified), Concentration: 0.2 mg/ml, Entrez Gene ID: 2048, Storage Buffer: Aqueous buffered solution containing ≤0.09% sodium azide.
Catalog No. NC3588611