Learn More
Cayman Chemical RBI 257maleate 10mg
SDP
Supplier:  Cayman Chemical 1722310 MG
Encompass_Preferred
A potent dopamine D4 receptor antagonist (Ki = 0.3 nM) that is selective over D2L, D3, D2-like, and D1 receptors by 1,721-, 439-, 1,336-, and 8,576-fold, respectively.{28464}
Catalog No. 50-296-5412