Learn More
TARGETMOL CHEMICALS INC RBIN-2 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T285065MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Rbin-2 is a cell-permeable potent selective and reversible inhibitor of Midasin (Mdn1).Rbin-2 directly targets the AAA+ ATPase Midasin and inhibits eukaryotic ribosome production making it a potent probe for the study of eukaryotic ribosome assembly processes. purity: 98%
Catalog No. 50-258-0508