Learn More
Bon Opus Biosciences Recombinant Human RNA-binding protein 20/RBM20(C-His) - Source: E. coli
SDP
Supplier:  Bon Opus Biosciences BP260450UG
Encompass_Preferred
Recombinant Human RNA-binding protein 20/RBM20 is produced by E.coli expression system and the target gene encoding E931-L1227 is expressed with a His tag at the C-terminus. Accession#Q5T481. Formulation: Supplied as a 0.2 um filtered solution of 500mM PBS, 300mM NaCl, 10%Glycerol, pH7.4
Catalog No. NC2733750