Learn More
Bon Opus Biosciences Recombinant Human RNA-binding protein 45 - Source: E. coli
SDP
Supplier:  Bon Opus Biosciences BP324250UG
Encompass_Preferred
Recombinant Human RNA-binding protein 45 is produced by E.coli expression system and the target gene encoding D2-S200 is expressed with a His tag at the C-terminus. Accession#Q8IUH3. Formulation: Supplied as a 0.2 um filtered solution of PBS, 300mM NaCl, 10%Glycerol, pH7.4
Catalog No. NC2733789