Learn More
Selleck Chemical LLC RBN-2397 5mg
SDP
Supplier:  Selleck Chemical LLC S89935MG
Encompass_Preferred
RBN-2397 is a potent, selective and orally active NAD+ competitive inhibitor PARP7 with IC50 of <3 nM and Kd of <0.001 ?M. RBN-2397 has the potential for the research of tumor treatment. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3466