Learn More
AdipoGen RBP4 human rec
SDP
Supplier:  AdipoGen AG40A0041C050
Encompass_Preferred
Protein. Human RBP4 aa 19-201 is fused at the N-terminus to a FLAG -tag. Source HEK 293 cells. Endotoxin content <0.1EU/µg purified protein LAL test Lonza. Liquid. 0.2µm-filtered solution in PBS, pH 7.2. Purity >90% SDS-PAGE. Retinol binding protein 4 RBP4 RBP is a 21kDa secreted protein, a member of the lipocalin family and is known as the primary transporter of retinol vitamin A to tissues. A recent report revealed RBP4 as an adipokine linking glucose transporter 4 GLUT4 suppression in adipose tissue to insulin. Elevated human and mouse serum RBP4 levels are associated with insulin resistance and its severity, obesity, and certain components of metabolic syndrome. Furthermore, human serum RBP4 levels are closely related to renal function.
Catalog No. 50-256-3045