Learn More
BioVendor, LLC RBP4 Mouse/Rat ELISA
SDP
Supplier:  BioVendor, LLC RAG007R
Encompass
Retinol binding protein (RBP) 4 is the only specific transport protein for vitamin A in the circulation whose function is to deliver vitamin to target tissues. In obesity and type 2 diabetes, expression of Glut4 is significantly impaired in adipocytes. Glucose transport via Glut4 is the rate-limiting step for glucose use by muscle and adipose tissue. Yang et al. noted that adipocyte-specific deletion of Gluts led to notable elevation of RBP4 causing systemic insulin resistance, and that reduction of RBP4 improved insulin resistance. This identified a novel role of RBP4 in regulating insulin action and RBP4 is recorded as an adipocyte-derived hormone. Thus, measurement of serum or plasma RBP4 is a useful means for understanding metabolic disorders. This sandwich ELISA (HRP-labelled antibody) quantitatively measures RBP4 in mouse/rat serum, urine, and cell culture supernatant. One kit (96 wells). Cal range: 0.188-12ng/ml. LoD: 60pg/ml. Other names:, PRBP, RBP, RBP4, PRO2222.
Catalog No. 50-274-3792