Learn More
Medchemexpress LLC RBP7 Human His 2ug

Supplier: Medchemexpress LLC HYP71251A2UG
RBP7 Protein, Human (His) is a recombinant human retinol-binding protein provided lyophilized for research applications. Expressed in E. coli and supplied with a His tag, it is intended for biochemical assays, ligand-binding studies, and antibody generation. The formulation and storage guidance support stable handling and long-term storage when aliquoted and frozen.
- Recombinant human RBP7 expressed in E. coli for consistent quality.
- His tag facilitates affinity purification and detection.
- Approximately 16 kDa molecular weight, compatible with SDS-PAGE analysis.
- Lyophilized formulation for extended shelf life and convenient reconstitution.
- Formulated in PBS with 300 mM NaCl, pH 7.4, to maintain stability.
- Low endotoxin level (<1 EU/μg) suitable for sensitive applications.
- Reconstitution recommended to ≥100 μg/mL; aliquot and freeze for long-term storage.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.