Learn More
Cayman Chemical RBromoenol lactoned7
SDP
Supplier:  Cayman Chemical 105341 MG
Encompass_Preferred
An internal standard for the quantification of (R)-BEL by GC- or LC-MS
Catalog No. 50-288-8217