Learn More
Aviva Systems Biology Corp Rcc2 Antibody - N-terminal region (ARP57289_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57289_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Rcc2. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 2610510H01Rik, 2610529N02Rik, AA536646, AA675016, Td60, mKIAA1470ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-292