Learn More
Abcam Recombinant human Hsp70 protein, 25UG
Supplier:  Abcam AB7843325UG
Encompass_Preferred
Recombinant human Hsp70 protein is a Human Full Length protein, in the 1 to 641 aa range, expressed in Escherichia coli, with >95% purity and suitable for ELISA, SDS-PAGE, WB, CBA, FuncS.
Protein/ Peptide/ Enzyme Synonyms: HSP72, HSPA1, HSX70, HSPA1A, Heat shock 70 kDa protein 1A, Heat shock 70 kDa protein 1, Heat shock protein family A member 1A, HSP70-1, HSP70.1
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1730178