Learn More
Medchemexpress LLC RCN3 Mouse HEK293 50ug | 50ug
SDP
Supplier:  Medchemexpress LLC HYP7717750UG
Encompass_Preferred
RCN3 Protein a probable molecular chaperone ensures pulmonary surfactant homeostasis by aiding in proper biosynthesis and transport of SP-A SP-D and ABCA3 It exhibits anti-fibrotic activity by negatively regulating type I and type III collagen secretion RCN3 transiently associates with immature PCSK6 suggesting its role in PCSK6 maturation and secretion RCN3 Protein Mouse (HEK293 His) is the recombinant mouse-derived RCN3 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-86453