Learn More
Electron Microscopy Sciences Veco Grid L2 x 0.6, Copper with 2000 x 600 µm Rectangle Slot
SDP
Supplier:  Electron Microscopy Sciences 0620CU
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Material: Copper
Slot: 2000 x 600 µm
Catalog No. 50-980-132