Learn More
ATCC RD; Rhabdomyosarcoma; Human (Homo sapiens)
Supplier:  ATCC CCL136
Encompass
Rhabdomyosarcoma (RD) cells are large, multinucleated, spindle-shaped cells isolated from muscle tissue. Use this cell line for your research and for the detection of viruses.
Catalog No. 50-238-1959
 Restricted Item