Learn More
Medchemexpress LLC RDR 02308 | 4155-82-2 | C19H15N3O3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1544385MG
Encompass_Preferred
RDR 02308 is a TLR4-MyD88 binding inhibitor that inhibits full-length β-lactamase, intended for research use only. In vitro studies show that RDR 02308 (0-10 μM; 1 h) inhibits LPS-mediated IL-6 production in RAW264.7 cells.
  • Inhibits TLR4-MyD88 binding.
  • Inhibits full-length beta-lactamase.
  • Intended for research use only.
  • Inhibits LPS-mediated IL-6 production in RAW264.7 cells.
  • Available in solid form.
  • Yellow to orange color.
  • Soluble in DMSO at 100 mg/mL.
  • Store powder at -20°C for up to 3 years or 4°C for up to 2 years.
  • Store in solvent at -80°C for up to 6 months or -20°C for up to 1 month.
Catalog No. 50-003-76539