Learn More
Medchemexpress LLC RdRP-IN-2 | 2863657-16-1 | 98.0% | 566.67 g·mol⁻¹ | C33H30N2O5S | 5 MG

Supplier: Medchemexpress LLC HY1394425MG
RdRP-IN-2 is a small-molecule research compound that inhibits RNA-dependent RNA polymerase (RdRp) and shows antiviral activity against SARS-CoV-2 in cell-based assays.
- Inhibits RdRp with reported IC50 of 41.2 μM.
- Demonstrates antiviral activity in Vero cells (EC50 = 527.3 nM).
- High chemical purity (98.0%).
- Molecular formula C33H30N2O5S; molar mass 566.67 g·mol⁻¹.
- Soluble in DMSO; offered as solid and as a 10 mM solution in DMSO.
- For research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.