Learn More
Amsbio LLC SV40 large T-antigen (GFP-Puro)/ EF1a lentiviral particles
Supplier:  Amsbio LLC LVP557GPPBS
Encompass
Pre-made lentiviral particles expressing SV40 large T-antigen under EF1a promoter/ containing GFP-puromycin fusion dual marker.
Catalog No. NC2735384