Learn More
STA PHARMACEUTICAL US LLC (R,E)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-4-(2,3-bis(tert-butoxycarbonyl)guanidino)butanoic acid | 1 g | CAS 1263047-29-5 | MDL MFCD13184917
Supplier: STA PHARMACEUTICAL US LLC STAAA122191GM
(R,E)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-4-(2,3-bis(tert-butoxycarbonyl)guanidino)butanoic acid is a Amino Acid reagent (Subcategory: Arg) sold by WuXi TIDES. Offered in 1 g. Store at 4 °C. SDS available for reference.
Specifications
- CAS: 1263047-29-5
- MDL: MFCD13184917
- InChIKey: FNEGEHCONNLNCC-HSZRJFAPSA-N
- Molecular Weight: 582.654
- Molecular Formula: C30H38N4O8
- Purity: ≥95%
- Container Type: 15 mL HDPE
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 7.8 g
- Commodity Code: 29252900
- Country Of Origin: China
- IUPAC: (R,Z)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-4-(2,3-bis(tert-butoxycarbonyl)guanidino)butanoic acid
- SMILES: CC(C)(OC(N/C(NCC[C@@H](NC(OCC1C2=C(C3=C1C=CC=C3)C=CC=C2)=O)C(O)=O)=N\C(OC(C)(C)C)=O)=O)C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.