Learn More
Chemrus Inc REACTION BLOCK FOR HOLDING MULTI-FLASKS
Supplier:  Chemrus Inc CR135014
Encompass
The anodized reaction block provides a safe and effective solution for replacing existing oil bath and heating mantles.  The block simultaneously holds 7 Liu Flasks (3 x 60 mL, 3 x 120 mL, 1 x 250 mL) and 6 vials (40 mL) to perform multi-flask reactions without the use of clamps.  The reaction block fits onto the circular top of the hot plate stirrer with a 135 mm diameter.  The wells provide space for inserting the thermometer and temperature sensor.   
Catalog No. NC0901014
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.