Learn More
Medchemexpress LLC Reactive blue 19 | 2580-78-1 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYD0331
Encompass_Preferred
Reactive Blue 19 is an anthraquinone dye primarily used in the textile industry for dyeing cellulose fibers. It is also utilized as a starting material for polymer dye production. While effective for dyeing, it is considered a toxic and undegradable organic pollutant that can pose risks to human health and aquatic life.
  • Used in textile industry
  • Applicable for dyeing cellulose fibers
  • Utilized in diagnostic assay manufacturing, hematology, and histology
  • Can serve as a starting material for polymer dyes
  • Known water pollutant, decolourization can be achieved via photodegradation
Catalog No. 50-004-14373