Learn More
Medchemexpress LLC Reactive Blue 19 | 2580-78-1 | 500 G
Supplier: Medchemexpress LLC HYD0331500G
Reactive Blue 19 is an anthraquinone dye utilized in the textile industry. It can also serve as a starting material for producing polymer dyes. This product is recognized as a toxic and undegradable organic pollutant, which poses a threat to human health and aquatic life.
- Used as an anthraquinone dye in the textile industry.
- Can be utilized as a starting material for the production of polymer dyes.