Learn More
Medchemexpress LLC HY-125815 500mg Medchemexpress, Reactive Blue 4 CAS:13324-20-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY125815500MG
Encompass_Preferred
Medchemexpress, HY-125815 500mg Reactive Blue 4 CAS:13324-20-4 Reactive Blue 4 is an anthraquinone dye, as a single colorimetric chemosensor for sequential determination of multiple analytes with different optical responses in aqueous media. Reactive Blue 4 is phytotoxic, cytotoxic and genotoxic. Reactive Blue 4 . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1084