Learn More
Ace Glass, Inc. 2000mL, one-piece jacketed pressure reactor with #25 center, (3)#15, (1)#7 side necks, 28/15 ball joint inlet/outlet, complete with addition funnel, condenser, pressure manifold and stirring assembly
SDP
Supplier:  Ace Glass, Inc. 6438250
Encompass
2000mL one-piece jacketed pressure reactor with a #25 center neck, (3) #15 and (1) #7 side necks, and a 28/15 o-ring ball joint on the inlet/outlet. Reactor system designed to perform low to moderate positive pressure reactions, synthesis and catalysis. Rated at 45psi at 100 degrees Centigrade. Supplied with addition funnel, condenser, pressure manifold, and stirring assembly.
Catalog No. 50-972-946
Add to Cart
Request a Quote