Learn More
Ace Glass, Inc. 500mL, one-piece jacketed pressure reactor with #25 center, (3)#15, (1)#7 side necks, 28/15 ball joint inlet/outlet, complete with addition funnel, condenser, pressure manifold and stirring assembly
SDP
Supplier:  Ace Glass, Inc. 6438240
Encompass
500mL one-piece jacketed pressure reactor with a #25 center neck, (3) #15 and (1) #7 side necks, and a 28/15 o-ring ball joint on the inlet/outlet. Reactor system designed to perform low to moderate positive pressure reactions, synthesis and catalysis. Rated at 45psi at 100 degrees Centigrade. Supplied with addition funnel, condenser, pressure manifold, and stirring assembly.
Catalog No. 50-978-721
Only null left
Add to Cart
Request a Quote