Learn More
Ace Glass, Inc. 1000mL, two-piece jacketed pressure reactor, complete with#11 Ace-Thred inlet/outlets, 100mm head with #15 center, (3)#15, (1)#7 side necks, addition funnel, condenser, pressure manifold and super magnet stirring assembly
SDP
Supplier:  Ace Glass, Inc. 6427468
Encompass
1000mL, two-piece jacketed pressure reactor, complete with#11 Ace-Thred inlet/outlets, 100mm head with #15 center, (3)#15, (1)#7 side necks, addition funnel, condenser, pressure manifold and super magnet stirring assembly
Catalog No. 50-972-905
Add to Cart
Request a Quote