Learn More
Chemglass Life Sciences Reactor Support Frame, 10L-20L, 19" W x 19" D x 66" H, 15.3" Valve Clearance
SDP
Supplier:  Chemglass Life Sciences CG1965X150
Encompass_Preferred
Reactor Support Frame, 10L-20L, 19" W x 19" D x 66" H, 15.3" Valve Clearance. Features:¿ Oversized Rugged 1" S.S.Pipe (1.315" OD) Uprights and Heavy Duty PTFE Coated Aluminum Fittings Provide Maximum Strength and Chemical Durability. ¿ All S.S.Uprights are Passivated to Inhibit Corrosion. ¿ Improved Support Clamp Auto-Centers Reactor and Lid, Holding it More Securely and Allowing Easier Installation/Removal of Glassware. ¿ Motor Mount for Electric or Air Motors. ¿ Open Air Design Provides Excellent Accessibility to Reactor and Glassware Components. ¿ Lower Support Plate has a ¿V¿ Shaped Opening that Allows an Unobstructed View of the Bottom Valve. ¿ Mobile: Large Swivel Casters with Locking Mechanisms.
Catalog No. 50-872-366