Learn More
Chemglass Life Sciences Chemglass 100 L Jacketed Process Reactor System, Squatty Cylindrical, 400 mm Flange, Complete
SDP
Supplier:  Chemglass Life Sciences CG1968MB9100
Encompass_Preferred
Complete 100 L jacketed glass reactor system for laboratory and pilot-scale process applications. The squatty, low-profile cylindrical vessel features a 400 mm ID flange, 7-neck lid with an 80 mm powder addition port, integrated agitation with a brushless DC motor and mechanical bearing, an included pressure relief manifold system, transparent safety shields, and mobile support with casters.
  • 100 L capacity
  • 400 mm ID flange
  • Jacketed glass vessel for temperature control
  • Brushless DC electric motor with mechanical bearing
  • 3/8 HP overhead motor, 30-500 RPM, 43 in-lbs
  • 7-neck lid with 80 mm powder addition port
  • 3 in beaded pipe zero-dead-space detachable drain valve
  • Pressure relief manifold system with insulated hoses and bulkhead fittings
  • Transparent safety shields on all sides with hinged front panel
  • Heavy duty 2 in diameter stainless-steel uprights and PTFE-coated aluminum fittings
Catalog No. NC3348044
Only null left
Add to Cart
Request a Quote