Learn More
Aat Bioquest ReadiLink Rapid iFluor 700 A
SDP
Supplier:  Aat Bioquest 1245
Encompass_Preferred
ReadiLink Rapid iFluor 700 Antibody Labeling Kit *Microscale Optimized for Labeling 50 μg Antibody Per Reaction*
Catalog No. 50-297-9314