Learn More
Waters Corp ReadyCal PS Kit, Solid, System Calibration, 4 pk
Supplier: Waters Corp 186007223
This Polystyrene Standards kit is designed for the APC System, offering a comprehensive set of calibration standards in solid form.
- Detection method: RI
- Formulation: Solid
- Storage condition: 4°C (Fridge)
- System type: APC
- Instrument: APC
- Shelf life: 36 months
- Shelf life upon reconstitution: 1 week
- Shipping condition: Ambient
- Mass: 2.25 mg
- Type: Calibration
- Application: System calibration
- Product type: Standards
- Units per package: 4 pk
- Includes black cap vial with polystyrene at MW 66K, 21.5K, 4.92K, 2.28K
- Includes blue cap vial with polystyrene at MW 44.2K, 15.7K, 3.47K, 1.25K
- Includes green cap vial with polystyrene at MW 28K, 9.13K
- Includes white cap vial with polystyrene at MW 34.8K, labeled as unknown
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.