Learn More
Regis Technologies Inc Pyridine, 2 x 25 ML
SDP
Supplier:  Regis Technologies Inc 1270013200
Encompass
Gas Chromatography (GC) Derivatization Reagent, Derivatization Grade Solvents. A high purity reagent packaged under nitrogen and sealed with Teflon-coated septa. This allows for easy access to sample without exposure to moisture and oxygen. Also known as azabenzene or azine.
Catalog No. 50-153-2989
 Restricted Item