Learn More
IMEB, Inc REAGENT ALCOHOL, 50%, 1 GALLON
SDP
Supplier:  IMEB, Inc TA11050
Encompass_Preferred
Tek-Select Reagent Alcohol 50%, 1 Gallon. Tek-Select Reagent Alcohol is composed of 90 parts ethyl alcohol, 5 parts methyl alcohol, and 5 parts isopropyl alcohol, v/v blend. Preferred mixture for dehydrating tissue samples prior to embedding and microscope slide staining. Tek-Select Reagent Alcohols are suitable for most histological procedures. 1 Gallon Bottle.
Catalog No. NC1897344